C12H316CE57F Chemcia Scientific, LLC

3-Chloro-pyridine 1-oxide

  • MDL number MFCD00128857
  • Cas Number : 1851-22-5
  • Molecular Weight : 129.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-chloropyridine 1-oxide
Inchi Code 1S/C5H4ClNO/c6-5-2-1-3-7(8)4-5/h1-4H
Inchi Key NEXUNCTUUDERGR-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1341

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1341
Availability details for
Quantity
  • Availability
  • Add to Cart