C12H316CE583 Chemcia Scientific, LLC

2,4-Difluoro-benzamidine

  • MDL number MFCD05662976
  • Cas Number : 885957-21-1
  • Molecular Weight : 156.13607

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Pale Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1345

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1345
Availability details for
Quantity
  • Availability
  • Add to Cart