C12H316CE584 Chemcia Scientific, LLC

2,6-Difluoro-benzamidine

  • MDL number MFCD05662968
  • Cas Number : 762229-49-2
  • Molecular Weight : 156.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,6-difluorobenzenecarboximidamide
Inchi Code 1S/C7H6F2N2/c8-4-2-1-3-5(9)6(4)7(10)11/h1-3H,(H3,10,11)
Inchi Key VCGXHAPBHZDDDJ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Brownish Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1346

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1346
Availability details for
Quantity
  • Availability
  • Add to Cart