C12H316CE585 Chemcia Scientific, LLC

(5-Chloro-3-fluoro-pyridin-2-yl)-hydrazine

  • MDL number MFCD09972183
  • Cas Number : 248255-70-1
  • Molecular Weight : 161.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-3-fluoro-2-hydrazinopyridine
Inchi Code 1S/C5H5ClFN3/c6-3-1-4(7)5(10-8)9-2-3/h1-2H,8H2,(H,9,10)
Inchi Key TZBBCECFLONWDG-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1347

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1347
Availability details for
Quantity
  • Availability
  • Add to Cart