C12H316CE587 Chemcia Scientific, LLC

2-Methoxy-5,6,7,8-tetrahydro-pyrido[4,3-d]pyrimidin hydrochloride

  • MDL number MFCD22570198
  • Cas Number : 1820619-84-8
  • Molecular Weight : 201.65734

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0~5
Physical Form Pale Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1199

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1199
Availability details for
Quantity
  • Availability
  • Add to Cart