C12H316CE588 Chemcia Scientific, LLC

Pyridin-4-yl-hydrazine

  • MDL number MFCD00731049
  • Cas Number : 27256-91-3
  • Molecular Weight : 109.13164

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1375

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1375
Availability details for
Quantity
  • Availability
  • Add to Cart