C12H316CE591 Chemcia Scientific, LLC

4-(2-Amino-phenyl)-4-oxo-butyric acid methyl ester

  • MDL number MFCD22565655
  • Cas Number : 57231-39-7
  • Molecular Weight : 207.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-(2-aminophenyl)-4-oxobutanoate
Inchi Code 1S/C11H13NO3/c1-15-11(14)7-6-10(13)8-4-2-3-5-9(8)12/h2-5H,6-7,12H2,1H3
Inchi Key LRIKFAUEEGOUQL-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1356

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1356
Availability details for
Quantity
  • Availability
  • Add to Cart