C12H316CE594 Chemcia Scientific, LLC

3-(1H-Indol-3-yl)-propionic acid methyl ester

  • MDL number MFCD00022778
  • Molecular Weight : 203.24291

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Orange Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1358

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1358
Availability details for
Quantity
  • Availability
  • Add to Cart