C12H316CE596 Chemcia Scientific, LLC

3-(1-Benzyl-1H-indol-3-yl)-propionic acid

  • MDL number MFCD03830273
  • Cas Number : 141071-79-6
  • Molecular Weight : 279.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5C
Physical Form Orange Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1378

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1378
Availability details for
Quantity
  • Availability
  • Add to Cart