C12H316CE5A3 Chemcia Scientific, LLC

2-tert-Butyl-5,7-dihydro-pyrrolo[3,4-d]pyrimidine-6-carboxylic acid tert-butyl ester

  • MDL number MFCD19981653
  • Cas Number : 1330754-01-2
  • Molecular Weight : 277.36946

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1363

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1363
Availability details for
Quantity
  • Availability
  • Add to Cart