C12H316CE5A6 Chemcia Scientific, LLC

3-Hydroxy-5,7-dihydro-pyrrolo[3,4-c]pyridazine-6-carboxylic acid benzyl ester

  • MDL number MFCD22565662
  • Cas Number : 1395493-21-6
  • Molecular Weight : 271.27801

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Gray Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1386

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1386
Availability details for
Quantity
  • Availability
  • Add to Cart