C12H316CE5B2 Chemcia Scientific, LLC

6-Chloro-imidazo[1,2-a]pyridine-2-carbaldehyde

  • MDL number MFCD06739239
  • Cas Number : 881841-30-1
  • Molecular Weight : 180.59485

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1397

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1397
Availability details for
Quantity
  • Availability
  • Add to Cart