C12H316CE5B3 Chemcia Scientific, LLC

3-Hydroxy-5-methyl-pyrazole-1-carboxylic acid tert-butyl ester

  • MDL number MFCD22565664
  • Cas Number : 915396-44-0
  • Molecular Weight : 198.22353

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1398

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1398
Availability details for
Quantity
  • Availability
  • Add to Cart