C12H316CE5C3 Chemcia Scientific, LLC

2,4,5,7-Tetrahydro-pyrazolo[3,4-c]pyridine-6-carboxylic acid tert-butyl ester

  • MDL number MFCD22381916
  • Cas Number : 871726-73-7
  • Molecular Weight : 223.27704

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1416

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1416
Availability details for
Quantity
  • Availability
  • Add to Cart