C12H316CE5C5 Chemcia Scientific, LLC

6-Iodo-nicotinic acid

  • MDL number MFCD03426924
  • Cas Number : 13054-02-9
  • Molecular Weight : 249.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-iodonicotinic acid
Inchi Code 1S/C6H4INO2/c7-5-2-1-4(3-8-5)6(9)10/h1-3H,(H,9,10)
Inchi Key PCGJGRDGMJRQAJ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1418

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1418
Availability details for
Quantity
  • Availability
  • Add to Cart