C12H316CE5CA Chemcia Scientific, LLC

[2-(Pyridin-2-yloxy)-ethyl]-carbamic acid tert-butyl ester

  • MDL number MFCD22565667
  • Cas Number : 1029715-22-7
  • Molecular Weight : 238.28886

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1423

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1423
Availability details for
Quantity
  • Availability
  • Add to Cart