C12H316CE5CE Chemcia Scientific, LLC

(2-Amino-4-trifluoromethyl-phenyl)-carbamic acid tert-butyl ester

  • MDL number MFCD09701248
  • Cas Number : 579474-48-9
  • Molecular Weight : 276.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 2-amino-4-(trifluoromethyl)phenylcarbamate
Inchi Code 1S/C12H15F3N2O2/c1-11(2,3)19-10(18)17-9-5-4-7(6-8(9)16)12(13,14)15/h4-6H,16H2,1-3H3,(H,17,18)
Inchi Key SAHCRZHMDWJEMU-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1447

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1447
Availability details for
Quantity
  • Availability
  • Add to Cart