C12H316CE5CF Chemcia Scientific, LLC

2-Bromo-6,7-dihydro-4H-pyrano[4,3-d]thiazole

  • MDL number MFCD22565668
  • Cas Number : 1093107-56-2
  • Molecular Weight : 220.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-6,7-dihydro-4H-pyrano[4,3-d]thiazole
Inchi Code 1S/C6H6BrNOS/c7-6-8-4-1-2-9-3-5(4)10-6/h1-3H2
Inchi Key WRFXNDSVRFCYJZ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1424

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1424
Availability details for
Quantity
  • Availability
  • Add to Cart