C12H316CE5D1 Chemcia Scientific, LLC

2-Amino-4,7-dihydro-5H-thieno[2,3-c]pyran-3-carbonitrile

  • MDL number MFCD00221038
  • Cas Number : 150986-82-6
  • Molecular Weight : 180.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-4,7-dihydro-5H-thieno[2,3-c]pyran-3-carbonitrile
Inchi Code 1S/C8H8N2OS/c9-3-6-5-1-2-11-4-7(5)12-8(6)10/h1-2,4,10H2
Inchi Key XCCSOSJCWRZLPA-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. 0~5
Physical Form Khaki Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1645

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1645
Availability details for
Quantity
  • Availability
  • Add to Cart