C12H316CE5DB Chemcia Scientific, LLC

5-Nitro-benzo[d]isoxazol-3-ylamine

  • MDL number MFCD02112721
  • Cas Number : 89793-82-8
  • Molecular Weight : 179.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-nitro-1,2-benzisoxazol-3-amine
Inchi Code 1S/C7H5N3O3/c8-7-5-3-4(10(11)12)1-2-6(5)13-9-7/h1-3H,(H2,8,9)
Inchi Key KIQGBLRPXXDRBZ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Orange Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1652

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1652
Availability details for
Quantity
  • Availability
  • Add to Cart