C12H316CE5DC Chemcia Scientific, LLC

5-Bromo-2-fluoro-benzamidine

  • MDL number MFCD09832178
  • Cas Number : 1100752-70-2
  • Molecular Weight : 217.04167

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1450

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1450
Availability details for
Quantity
  • Availability
  • Add to Cart