C12H316CE5DF Chemcia Scientific, LLC

2,4,6,7-Tetrahydro-pyrazolo[4,3-c]pyridine-5-carboxylic acid benzyl ester

  • MDL number MFCD18074543
  • Cas Number : 1355171-29-7
  • Molecular Weight : 257.29455

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1453

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1453
Availability details for
Quantity
  • Availability
  • Add to Cart