C12H316CE5EA Chemcia Scientific, LLC

1,2,3,4-Tetrahydro-quinolin-7-ylamine

  • MDL number MFCD08063884
  • Cas Number : 153856-89-4
  • Molecular Weight : 148.20939

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form Black Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1639

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1639
Availability details for
Quantity
  • Availability
  • Add to Cart