C12H316CE5EF Chemcia Scientific, LLC

1-Bromo-5-methoxy-2,4-dinitro-benzene

  • MDL number MFCD00624873
  • Cas Number : 181995-71-1
  • Molecular Weight : 277.0323

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1676

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1676
Availability details for
Quantity
  • Availability
  • Add to Cart