C12H316CE5F1 Chemcia Scientific, LLC

2-Bromo-4-methoxy-1,3-dinitro-benzene

  • MDL number MFCD23115358
  • Cas Number : 1421312-12-0
  • Molecular Weight : 277.0323

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1677

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1677
Availability details for
Quantity
  • Availability
  • Add to Cart