C12H316CE60A Chemcia Scientific, LLC

Ethyl [(5-chloro-2-pyridinyl)amino]carbothioylcarbamate

  • MDL number MFCD01833204
  • Cas Number : 78852-50-3
  • Molecular Weight : 259.71595

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1657

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1657
Availability details for
Quantity
  • Availability
  • Add to Cart