C12H316CE60D Chemcia Scientific, LLC

1-Nitroso-2,3-dihydro-1H-indole

  • MDL number MFCD01110239
  • Cas Number : 7633-57-0
  • Molecular Weight : 148.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-nitrosoindoline
Inchi Code 1S/C8H8N2O/c11-9-10-6-5-7-3-1-2-4-8(7)10/h1-4H,5-6H2
Inchi Key QVXPAHMYCSEIMA-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Beige Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1661

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1661
Availability details for
Quantity
  • Availability
  • Add to Cart