C12H316CE610 Chemcia Scientific, LLC

(2,3-Dihydro-indol-1-yl)-acetic acid methyl ester

  • MDL number MFCD05147568
  • Cas Number : 39597-68-7
  • Molecular Weight : 191.23176

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Black Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1683

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1683
Availability details for
Quantity
  • Availability
  • Add to Cart