C12H316CE613 Chemcia Scientific, LLC

2-Amino-1-(4-methoxy-phenyl)-ethanol

  • MDL number MFCD03838266
  • Cas Number : 55275-61-1
  • Molecular Weight : 167.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-1-(4-methoxyphenyl)ethanol
Inchi Code 1S/C9H13NO2/c1-12-8-4-2-7(3-5-8)9(11)6-10/h2-5,9,11H,6,10H2,1H3
Inchi Key LQEPONWCAMNCOY-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1691

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1691
Availability details for
Quantity
  • Availability
  • Add to Cart