C12H316CE615 Chemcia Scientific, LLC

1-(3-Methylamino-phenyl)-ethanone

  • MDL number MFCD16470104
  • Cas Number : 42865-75-8
  • Molecular Weight : 149.19412

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1693

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1693
Availability details for
Quantity
  • Availability
  • Add to Cart