C12H316CE61B Chemcia Scientific, LLC

(3-Bromo-4-oxo-cyclohexyl)-carbamic acid tert-butyl ester

  • MDL number MFCD23115355
  • Cas Number : 1073632-93-5
  • Molecular Weight : 292.17501

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1700

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1700
Availability details for
Quantity
  • Availability
  • Add to Cart