C12H316CE61C Chemcia Scientific, LLC

(2-Amino-4,5,6,7-tetrahydro-benzothiazol-6-yl)-carbamic acid tert-butyl ester

  • MDL number MFCD13195378
  • Cas Number : 820231-27-4
  • Molecular Weight : 269.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 2-amino-4,5,6,7-tetrahydro-1,3-benzothiazol-6-ylcarbamate
Inchi Code 1S/C12H19N3O2S/c1-12(2,3)17-11(16)14-7-4-5-8-9(6-7)18-10(13)15-8/h7H,4-6H2,1-3H3,(H2,13,15)(H,14,16)
Inchi Key CBTULJANIYAMDM-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1705

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1705
Availability details for
Quantity
  • Availability
  • Add to Cart