C12H316CE621 Chemcia Scientific, LLC

3-Hydroxy-7,8-dihydro-5H-pyrido[4,3-c]pyridazine-6-carboxylic acid benzyl ester

  • MDL number MFCD23115356
  • Cas Number : 1354963-19-1
  • Molecular Weight : 285.3051

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1710

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1710
Availability details for
Quantity
  • Availability
  • Add to Cart