C12H316CE622 Chemcia Scientific, LLC

(2,6-Dichloro-phenyl)-methyl-amine hydrochloride

  • MDL number MFCD22421827
  • Cas Number : 1423029-60-0
  • Molecular Weight : 212.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,6-dichloro-N-methylaniline hydrochloride
Inchi Code 1S/C7H7Cl2N.ClH/c1-10-7-5(8)3-2-4-6(7)9;/h2-4,10H,1H3;1H
Inchi Key IPYMEACBPVDBCS-UHFFFAOYSA-N

Description

Purity 98%
Salt Data HCl
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1718

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1718
Availability details for
Quantity
  • Availability
  • Add to Cart