C12H316CE624 Chemcia Scientific, LLC

Methyl-(4-methyl-2-nitro-phenyl)-amine

  • MDL number MFCD07437778
  • Molecular Weight : 166.1811

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Orange-Red Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1707

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1707
Availability details for
Quantity
  • Availability
  • Add to Cart