C12H316CE626 Chemcia Scientific, LLC

5-Amino-4-chloro-2-methoxy-benzoic acid methyl ester hydrochloride

  • MDL number MFCD26792539
  • Cas Number : 1951445-00-3
  • Molecular Weight : 252.09892

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Salt Data HCl
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1714

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1714
Availability details for
Quantity
  • Availability
  • Add to Cart