C12H316CE627 Chemcia Scientific, LLC

2-Amino-4-iodo-phenol

  • MDL number MFCD14705890
  • Cas Number : 99969-17-2
  • Molecular Weight : 235.02522

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1711

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1711
Availability details for
Quantity
  • Availability
  • Add to Cart