C12H316CE628 Chemcia Scientific, LLC

6-Chloro-4-formyl-nicotinic acid hydrate

  • MDL number MFCD18258739
  • Cas Number : 1031433-06-3
  • Molecular Weight : 203.58317

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1729

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1729
Availability details for
Quantity
  • Availability
  • Add to Cart