C12H316CE62D Chemcia Scientific, LLC

6-Iodo-nicotinic acid methyl ester

  • MDL number MFCD12546492
  • Cas Number : 173157-33-0
  • Molecular Weight : 263.03577

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1732

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1732
Availability details for
Quantity
  • Availability
  • Add to Cart