C12H316CE62E Chemcia Scientific, LLC

4-Hydroxy-3-iodo-benzoic acid

  • MDL number MFCD00234263
  • Cas Number : 37470-46-5
  • Molecular Weight : 264.0205

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1773

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1773
Availability details for
Quantity
  • Availability
  • Add to Cart