C12H316CE62F Chemcia Scientific, LLC

3-Bromo-N-methylpyridin-2-amine

  • MDL number MFCD07374349
  • Cas Number : 214977-38-5
  • Molecular Weight : 187.04009

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5C
Shipping Temperature Ambient
Country of Origin USA
Product Link http://www.chemcia.com/product_fl02.asp?id=1719

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1719
Availability details for
Quantity
  • Availability
  • Add to Cart