C12H316CE633 Chemcia Scientific, LLC

3-Methyl-4-nitro-pyrazole-1-carboxylic acid tert-butyl ester

  • MDL number MFCD23115360
  • Cas Number : 847139-22-4
  • Molecular Weight : 227.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-methyl-4-nitro-1H-pyrazole-1-carboxylate
Inchi Code 1S/C9H13N3O4/c1-6-7(12(14)15)5-11(10-6)8(13)16-9(2,3)4/h5H,1-4H3
Inchi Key BDFNCVSBBDMLAH-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1725

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1725
Availability details for
Quantity
  • Availability
  • Add to Cart