C12H316CE634 Chemcia Scientific, LLC

4-Amino-3-methyl-pyrazole-1-carboxylic acid tert-butyl ester

  • MDL number MFCD23115361
  • Cas Number : 847139-23-5
  • Molecular Weight : 197.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 4-amino-3-methyl-1H-pyrazole-1-carboxylate
Inchi Code 1S/C9H15N3O2/c1-6-7(10)5-12(11-6)8(13)14-9(2,3)4/h5H,10H2,1-4H3
Inchi Key AZEOFHCJPAQKHH-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1726

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1726
Availability details for
Quantity
  • Availability
  • Add to Cart