C12H316CE63A Chemcia Scientific, LLC

Pyrrolo[2,3-b]pyridin-1-yl-acetic acid methyl ester

  • Cas Number : 172647-94-8
  • Molecular Weight : 190.2034

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form Orange Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1734

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1734
Availability details for
Quantity
  • Availability
  • Add to Cart