C12H316CE63C Chemcia Scientific, LLC

8-Iodo-6-methyl-imidazo[1,2-a]pyridine-2-carboxylic acid ethyl ester hydrobromide

  • Cas Number : 1951441-86-3
  • Molecular Weight : 411.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 8-iodo-6-methylimidazo[1,2-a]pyridine-2-carboxylate hydrobromide
Inchi Code 1S/C11H11IN2O2.BrH/c1-3-16-11(15)9-6-14-5-7(2)4-8(12)10(14)13-9;/h4-6H,3H2,1-2H3;1H
Inchi Key BXSXCCURXYEBND-UHFFFAOYSA-N

Description

Purity 95%
Salt Data HBr
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1736

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1736
Availability details for
Quantity
  • Availability
  • Add to Cart