C12H316CE645 Chemcia Scientific, LLC

5-Iodo-2,6-dimethyl-1H-pyrimidin-4-one

  • MDL number MFCD23115364
  • Cas Number : 83410-37-1
  • Molecular Weight : 250.03989

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1770

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1770
Availability details for
Quantity
  • Availability
  • Add to Cart