C12H316CE649 Chemcia Scientific, LLC

5-Bromo-2,6-dimethyl-pyrimidin-4-ol

  • MDL number MFCD16620430
  • Molecular Weight : 203.03949

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1749

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1749
Availability details for
Quantity
  • Availability
  • Add to Cart