C12H316CE64E Chemcia Scientific, LLC

2-Amino-5-bromo-4-chloro-benzoic acid methyl ester

  • MDL number MFCD08752643
  • Cas Number : 765211-09-4
  • Molecular Weight : 264.50749

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1757

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1757
Availability details for
Quantity
  • Availability
  • Add to Cart