C12H316CE651 Chemcia Scientific, LLC

(5-Fluoro-pyridin-2-yl)-carbamic acid tert-butyl ester

  • Cas Number : 1260794-50-0
  • Molecular Weight : 212.22571

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1762

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1762
Availability details for
Quantity
  • Availability
  • Add to Cart