C12H316CE652 Chemcia Scientific, LLC

3-Chloromethyl-1H-pyrazole-4-carboxylic acid ethyl ester

  • MDL number MFCD07357430
  • Cas Number : 137487-60-6
  • Molecular Weight : 188.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 3-(chloromethyl)-1H-pyrazole-4-carboxylate
Inchi Code 1S/C7H9ClN2O2/c1-2-12-7(11)5-4-9-10-6(5)3-8/h4H,2-3H2,1H3,(H,9,10)
Inchi Key SJOBUQJQZHZTPG-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2175

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2175
Availability details for
Quantity
  • Availability
  • Add to Cart